missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-Boc-S-(4-methoxybenzyl)-D-cysteine, 95%

Brand:  Alfa Aesar™ H62302.MD

Additional Details : CAS Number : 58290-35-0 Weight : 0.00025kg

Quantity 250mg

 View more versions of this product

Product Code. 15458838

  • 36.50 EUR / 250mg


Estimated Shipment date
from Supplier 23-04-2019
Add to basket

Specifications

Chemical Identifiers

58290-35-0
341.422
VRTXRNJMNFVTOM-CYBMUJFWSA-N
6992555
CC(C)(C)OC(=O)NC(CSCC1=CC=C(C=C1)OC)C(=O)O
C16H23NO5S
MFCD00076922
boc-s-4-methoxybenzyl-d-cysteine, boc-d-cys pmeobzl-oh, boc-d-cys 4-mob-oh, boc-d-cys 4-meo-bzl-oh, boc-d-cys mob-oh, boc-s-p-methoxybenzyl-d-cysteine, s-2-tert-butoxycarbonyl amino-3-4-methoxybenzyl thio propanoic acid, 2s-2-tert-butoxycarbonyl amino-3-4-methoxyphenyl methyl sulfanyl propanoic acid, ambotzbaa5430, n-tert-butyloxycarbonyl-s-p-methoxybenzyl-d-cysteine
(2S)-3-[(4-methoxyphenyl)methylsulfanyl]-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid

Specifications

N-Boc-S-(4-methoxybenzyl)-D-cysteine
C16H23NO5S
boc-s-4-methoxybenzyl-d-cysteine, boc-d-cys pmeobzl-oh, boc-d-cys 4-mob-oh, boc-d-cys 4-meo-bzl-oh, boc-d-cys mob-oh, boc-s-p-methoxybenzyl-d-cysteine, s-2-tert-butoxycarbonyl amino-3-4-methoxybenzyl thio propanoic acid, 2s-2-tert-butoxycarbonyl amino-3-4-methoxyphenyl methyl sulfanyl propanoic acid, ambotzbaa5430, n-tert-butyloxycarbonyl-s-p-methoxybenzyl-d-cysteine
CC(C)(C)OC(=O)NC(CSCC1=CC=C(C=C1)OC)C(=O)O
341.422
341.42
95%
58290-35-0
MFCD00076922
VRTXRNJMNFVTOM-CYBMUJFWSA-N
(2S)-3-[(4-methoxyphenyl)methylsulfanyl]-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid
6992555
Solid
250mg
Videos
SDS