missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-Boc-N-methyl-D-alanine, 98%

Brand:  Alfa Aesar™ H62510.MD

Additional Details : CAS Number : 19914-38-6 Weight : 0.00025kg

Quantity 250mg

 View more versions of this product

Product Code. 15459038

  • 60.00 EUR / 250mg


Estimated Shipment date
from Supplier 24-04-2019
Add to basket

Specifications

Chemical Identifiers

19914-38-6
203.238
VLHQXRIIQSTJCQ-ZCFIWIBFSA-N
16211306
CC(C(=O)O)N(C)C(=O)OC(C)(C)C
C9H17NO4
MFCD00063137
boc-n-methyl-d-alanine, boc-n-me-d-ala-oh, n-boc-n-methyl-d-alanine, boc-d-n-me-ala-oh, boc-n-alpha-methyl-d-alanine, r-2-tert-butoxycarbonyl methyl amino propanoic acid, n-tert-butoxycarbonyl-n-methyl-d-alanine, r-2-tert-butoxycarbonyl methyl amino propanoic acid, d-alanine, n-1,1-dimethylethoxy carbonyl-n-methyl, 2r-2-tert-butoxycarbonyl methyl amino propanoic acid
(2R)-2-[methyl-[(2-methylpropan-2-yl)oxycarbonyl]amino]propanoic acid

Specifications

N-Boc-N-methyl-D-alanine
C9H17NO4
3606540
VLHQXRIIQSTJCQ-ZCFIWIBFSA-N
(2R)-2-[methyl-[(2-methylpropan-2-yl)oxycarbonyl]amino]propanoic acid
16211306
Powder
White
250mg
19914-38-6
MFCD00063137
boc-n-methyl-d-alanine, boc-n-me-d-ala-oh, n-boc-n-methyl-d-alanine, boc-d-n-me-ala-oh, boc-n-alpha-methyl-d-alanine, r-2-tert-butoxycarbonyl methyl amino propanoic acid, n-tert-butoxycarbonyl-n-methyl-d-alanine, r-2-tert-butoxycarbonyl methyl amino propanoic acid, d-alanine, n-1,1-dimethylethoxy carbonyl-n-methyl, 2r-2-tert-butoxycarbonyl methyl amino propanoic acid
CC(C(=O)O)N(C)C(=O)OC(C)(C)C
203.238
203.24
98%
90°C to 94°C
Videos
SDS