missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Boc-L-isoleucine, 98+%, Thermo Scientific Chemicals
missing translation for 'orderingAttributeHoverText'
Quantity:
1 g
5 g
25 g
Beschreibung
N-Boc-L-isoleucine is used to prepare BOC-L-isoleucine hydroxysuccinimide ester by using 1-hydroxybenzotriazole and 1-ethyl-3-<3-(dimethylamino)propyl>carbodiimide hydrochloride.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
N-Boc-L-isoleucine is used to prepare BOC-L-isoleucine hydroxysuccinimide ester by using 1-hydroxybenzotriazole and 1-ethyl-3-<3-(dimethylamino)propyl>carbodiimide hydrochloride.
Solubility
Soluble in methanol. Insoluble in water.
Notes
Incompatible with strong oxidizing agents, strong acids and strong bases. Store in a cool place.
N-Boc-L-isoleucine is used to prepare BOC-L-isoleucine hydroxysuccinimide ester by using 1-hydroxybenzotriazole and 1-ethyl-3-<3-(dimethylamino)propyl>carbodiimide hydrochloride.
Solubility
Soluble in methanol. Insoluble in water.
Notes
Incompatible with strong oxidizing agents, strong acids and strong bases. Store in a cool place.
Spezifikation
Melting Point | 67°C to 70°C |
Quantity | 1 g |
Beilstein | 1711700 |
Solubility Information | Soluble in methanol. Insoluble in water. |
Formula Weight | 231.06 |
Percent Purity | ≥98% |
Chemical Name or Material | N-Boc-L-isoleucine |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'