missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-Boc-L-alanyl-L-alanine methyl ester, 97%

Brand:  Alfa Aesar™ H62550.03

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15459068

  • 201.00 EUR / 1g


Estimated Shipment date
from Supplier 19-02-2019
Add to basket

Specifications

Chemical Identifiers

19794-10-6
274.317
KRWPOGQDZNCNLK-YUMQZZPRSA-N
10671868
CC(C(=O)NC(C)C(=O)OC)NC(=O)OC(C)(C)C
C12H22N2O5
MFCD00235758
boc-ala-ala-ome, methyl 2s-2-2s-2-tert-butoxy carbonyl amino propanamido propanoate, boc-l-ala-l-ala-ome, boc-l-alanyl-l-alanine methylester, s-methyl 2-s-2-boc-amino propanamido propionate, methyl 2s-2-2s-2-tert-butoxycarbonyl amino propanoyl amino propanoate, methyl 2s-2-2s-2-tert-butoxycarbonyl amino propanamido propanoate
methyl (2S)-2-[[(2S)-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoyl]amino]propanoate

Specifications

N-Boc-L-alanyl-L-alanine methyl ester
C12H22N2O5
boc-ala-ala-ome, methyl 2s-2-2s-2-tert-butoxy carbonyl amino propanamido propanoate, boc-l-ala-l-ala-ome, boc-l-alanyl-l-alanine methylester, s-methyl 2-s-2-boc-amino propanamido propionate, methyl 2s-2-2s-2-tert-butoxycarbonyl amino propanoyl amino propanoate, methyl 2s-2-2s-2-tert-butoxycarbonyl amino propanamido propanoate
CC(C(=O)NC(C)C(=O)OC)NC(=O)OC(C)(C)C
274.317
274.31
97%
19794-10-6
MFCD00235758
KRWPOGQDZNCNLK-YUMQZZPRSA-N
methyl (2S)-2-[[(2S)-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoyl]amino]propanoate
10671868
Solid
1g
Videos
SDS