missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-Boc-D-aspartic acid, 98%

Brand:  Alfa Aesar™ H62820.06

Additional Details : CAS Number : 62396-48-9 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15459288

  • 49.00 EUR / 5g


Estimated Shipment date
from Supplier 18-06-2019
Add to basket

Specifications

Chemical Identifiers

62396-48-9
233.22
KAJBMCZQVSQJDE-RXMQYKEDSA-N
7021110
CC(C)(C)OC(=O)NC(CC(=O)O)C(=O)O
C9H15NO6
MFCD00798618
boc-d-aspartic acid, boc-d-asp-oh, n-boc-d-aspartic acid, n-tert-butoxycarbonyl-d-aspartic acid, d-aspartic acid, n-1,1-dimethylethoxy carbonyl, 2r-2-tert-butoxycarbonyl amino butanedioic acid, boc-d-asparticacid, pubchem12159, n-t-boc-d-aspartic acid, ksc494c3p
(2R)-2-[(2-methylpropan-2-yl)oxycarbonylamino]butanedioic acid

Specifications

N-Boc-D-aspartic acid
C9H15NO6
boc-d-aspartic acid, boc-d-asp-oh, n-boc-d-aspartic acid, n-tert-butoxycarbonyl-d-aspartic acid, d-aspartic acid, n-1,1-dimethylethoxy carbonyl, 2r-2-tert-butoxycarbonyl amino butanedioic acid, boc-d-asparticacid, pubchem12159, n-t-boc-d-aspartic acid, ksc494c3p
CC(C)(C)OC(=O)NC(CC(=O)O)C(=O)O
233.22
233.22
98%
62396-48-9
MFCD00798618
KAJBMCZQVSQJDE-RXMQYKEDSA-N
(2R)-2-[(2-methylpropan-2-yl)oxycarbonylamino]butanedioic acid
7021110
Solid
5g
Videos
SDS