missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-Boc-2,4,5-trifluoro-D-phenylalanine, 98%

Brand:  Alfa Aesar™ H62878.MD

Additional Details : CAS Number : 486460-09-7 Weight : 0.00025kg

Quantity 250mg

 View more versions of this product

Product Code. 15459328

  • 63.20 EUR / 250mg


Estimated Shipment date
from Supplier 24-04-2019
Add to basket

Specifications

Chemical Identifiers

486460-09-7
319.28
FTWHJNNFVISGNQ-LLVKDONJSA-N
7146200
CC(C)(C)OC(=O)NC(CC1=CC(=C(C=C1F)F)F)C(=O)O
C14H16F3NO4
MFCD06659118
r-2-tert-butoxycarbonyl amino-3-2,4,5-trifluorophenyl propanoic acid, boc-2,4,5-trifluoro-d-phenylalanine, 2r-2-2-methylpropan-2-yl oxycarbonylamino-3-2,4,5-trifluorophenyl propanoic acid, 2r-2-tert-butoxycarbonyl amino-3-2,4,5-trifluorophenyl propanoic acid, boc-d-2-amino-3-2,4,5-trifluoro-phenyl alanine, r-n-tert-butoxycarbonyl-2,4,5-trifluorophenylalanine, r-2-tert-butoxycarbonyl amino-3-2,4,5-trifluorophenyl propanoicacid
(2R)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-3-(2,4,5-trifluorophenyl)propanoic acid

Specifications

N-Boc-2,4,5-trifluoro-D-phenylalanine
C14H16F3NO4
r-2-tert-butoxycarbonyl amino-3-2,4,5-trifluorophenyl propanoic acid, boc-2,4,5-trifluoro-d-phenylalanine, 2r-2-2-methylpropan-2-yl oxycarbonylamino-3-2,4,5-trifluorophenyl propanoic acid, 2r-2-tert-butoxycarbonyl amino-3-2,4,5-trifluorophenyl propanoic acid, boc-d-2-amino-3-2,4,5-trifluoro-phenyl alanine, r-n-tert-butoxycarbonyl-2,4,5-trifluorophenylalanine, r-2-tert-butoxycarbonyl amino-3-2,4,5-trifluorophenyl propanoicacid
CC(C)(C)OC(=O)NC(CC1=CC(=C(C=C1F)F)F)C(=O)O
319.28
319.28
98%
486460-09-7
MFCD06659118
FTWHJNNFVISGNQ-LLVKDONJSA-N
(2R)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-3-(2,4,5-trifluorophenyl)propanoic acid
7146200
Solid
250mg
Videos
SDS