missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-Benzyloxycarbonyl-1-Boc-L-tryptophan dicyclohexylammonium salt, 98%

Brand:  Alfa Aesar™ H62015.06

Additional Details : CAS Number : 218938-57-9 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15446926

  • 268.00 EUR / 5g


Estimated Shipment date
from Supplier 21-10-2019
Add to basket

Specifications

Chemical Identifiers

218938-57-9
619.803
BUQVAJPBCWSLMH-FYZYNONXSA-N
16218811
CC(C)(C)OC(=O)N1C=C(C2=CC=CC=C21)CC(C(=O)O)NC(=O)OCC3=CC=CC=C3.C1CCC(CC1)NC2CCCCC2
C36H49N3O6
MFCD01318754
z-trp boc-oh.dcha, z-trp boc-oh dcha, z-l-trp boc-oh, z-trp boc-ohcha, z-trp boc-oh dicyclohexylammonium salt, 1-boc-nalpha-cbz-l-tryptophan dicyclohexylamine salt, n, a-z-n in-boc-l-tryptophan dicyclohexylammonium salt, 2s-2-benzyloxy carbonyl amino-3-1-tert-butoxycarbonyl indol-3-yl propanoic acid; dicha
N-cyclohexylcyclohexanamine;(2S)-3-[1-[(2-methylpropan-2-yl)oxycarbonyl]indol-3-yl]-2-(phenylmethoxycarbonylamino)propanoic acid

Specifications

N-Benzyloxycarbonyl-1-Boc-L-tryptophan dicyclohexylammonium salt
C36H49N3O6
z-trp boc-oh.dcha, z-trp boc-oh dcha, z-l-trp boc-oh, z-trp boc-ohcha, z-trp boc-oh dicyclohexylammonium salt, 1-boc-nalpha-cbz-l-tryptophan dicyclohexylamine salt, n, a-z-n in-boc-l-tryptophan dicyclohexylammonium salt, 2s-2-benzyloxy carbonyl amino-3-1-tert-butoxycarbonyl indol-3-yl propanoic acid; dicha
CC(C)(C)OC(=O)N1C=C(C2=CC=CC=C21)CC(C(=O)O)NC(=O)OCC3=CC=CC=C3.C1CCC(CC1)NC2CCCCC2
619.803
619.79
98%
218938-57-9
MFCD01318754
BUQVAJPBCWSLMH-FYZYNONXSA-N
N-cyclohexylcyclohexanamine;(2S)-3-[1-[(2-methylpropan-2-yl)oxycarbonyl]indol-3-yl]-2-(phenylmethoxycarbonylamino)propanoic acid
16218811
Solid
5g
Videos
SDS Documents