missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-Benzyl-N,N-bis(2-chloroethyl)amine hydrochloride, 95%

Brand:  Alfa Aesar™ H64057.MD

Additional Details : CAS Number : 10429-82-0 Weight : 0.00025kg Transport : UN number : 2923 Chem class : 8 Pack group : II

Quantity 250mg

 View more versions of this product

Product Code. 15458676

  • 26.20 EUR / 250mg


Estimated Shipment date
from Supplier 19-02-2019
Add to basket

Specifications

Chemical Identifiers

10429-82-0
268.606
AZRWNJFEUSHORT-UHFFFAOYSA-N
2777881
C1=CC=C(C=C1)CN(CCCl)CCCl.Cl
C11H16Cl3N
MFCD00185654
n-benzyl-2-chloro-n-2-chloroethyl ethanamine hydrochloride, n-benzyl-bis 2-chloroethyl amine hydrochloride, benzylbis 2-chloroethyl amine hydrochloride, n-benzyl-n,n-bis 2-chloroethyl amine hydrochloride, unii-yfi3592jvs, n-benzyl-bis 2-chloroethyl amine hcl, benzenemethanamine, n,n-bis 2-chloroethyl-, hydrochloride, n-benzyl-2-chloro-n-2-chloroethyl ethan-1-amine hydrochloride, yfi3592jvs, ksc913g7f
N-benzyl-2-chloro-N-(2-chloroethyl)ethanamine;hydrochloride

Specifications

N-Benzyl-N,N-bis(2-chloroethyl)amine hydrochloride
147°C to 150°C
10429-82-0
MFCD00185654
3916139
AZRWNJFEUSHORT-UHFFFAOYSA-N
N-benzyl-2-chloro-N-(2-chloroethyl)ethanamine;hydrochloride
2777881
Powder
White
250mg
C11H16Cl3N
UN2923
n-benzyl-2-chloro-n-2-chloroethyl ethanamine hydrochloride, n-benzyl-bis 2-chloroethyl amine hydrochloride, benzylbis 2-chloroethyl amine hydrochloride, n-benzyl-n,n-bis 2-chloroethyl amine hydrochloride, unii-yfi3592jvs, n-benzyl-bis 2-chloroethyl amine hcl, benzenemethanamine, n,n-bis 2-chloroethyl-, hydrochloride, n-benzyl-2-chloro-n-2-chloroethyl ethan-1-amine hydrochloride, yfi3592jvs, ksc913g7f
C1=CC=C(C=C1)CN(CCCl)CCCl.Cl
268.606
268.61
95%
Videos
SDS