missing translation for 'onlineSavingsMsg'
Learn More

N-Benzyl-N-ethyl-2,6-difluorobenzamide, 97%, Thermo Scientific™

Catalog Number p-7039914
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
1421263-95-7
C16H15F2NO
275.299
MFCD22683068
WWMNXFJMLWOKHV-UHFFFAOYSA-N
86975988
N-benzyl-N-ethyl-2,6-difluorobenzamide
CCN(CC1=CC=CC=C1)C(=O)C2=C(C=CC=C2F)F
This item is not returnable. View return policy
1421263-95-7
C16H15F2NO
275.299
MFCD22683068
WWMNXFJMLWOKHV-UHFFFAOYSA-N
86975988
N-benzyl-N-ethyl-2,6-difluorobenzamide
CCN(CC1=CC=CC=C1)C(=O)C2=C(C=CC=C2F)F

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 250 mg
Formula Weight 275.29
Percent Purity 97%
Chemical Name or Material N-Benzyl-N-ethyl-2,6-difluorobenzamide
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'