Learn More
N-Benzyl-4-methoxybenzamide, 97%, Thermo Scientific™
Catalog Number p-7053238
Click to view available options
Quantity:
250 mg
1 g
7465-87-4
C15H15NO2
241.29
MFCD00507374
GEFZSLMGZREDTO-UHFFFAOYSA-N
n-benzyl-4-methoxy-benzamide, aronis25526, 4-methoxyphenyl-n-benzylcarboxamide
346035
N-benzyl-4-methoxybenzamide
COC1=CC=C(C=C1)C(=O)NCC2=CC=CC=C2
7465-87-4
C15H15NO2
241.29
MFCD00507374
GEFZSLMGZREDTO-UHFFFAOYSA-N
n-benzyl-4-methoxy-benzamide, aronis25526, 4-methoxyphenyl-n-benzylcarboxamide
346035
N-benzyl-4-methoxybenzamide
COC1=CC=C(C=C1)C(=O)NCC2=CC=CC=C2
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Solubility
Soluble in water.
Notes
Keep container tightly closed. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY
Quantity 250 mg
Solubility Information Soluble in water.
Formula Weight 241.29
Percent Purity 97%
Physical Form Solid
Chemical Name or Material N-Benzyl-4-methoxybenzamide