missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-(4-Ethoxyphenyl)-2-methoxybenzamide, 97%

Brand:  Alfa Aesar™ H59767.03

Additional Details : CAS Number : 306279-64-1 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15486168

  • 646.00 EUR / 1g


Estimated Shipment date
from Supplier 31-08-2018
Add to basket

Specifications

Chemical Identifiers

306279-64-1
271.316
YOBHVPWWYDUPFT-UHFFFAOYSA-N
C16H17NO3
MFCD00841328
700573

Specifications

N-(4-Ethoxyphenyl)-2-methoxybenzamide
97%
MFCD00841328
YOBHVPWWYDUPFT-UHFFFAOYSA-N
N-(4-ethoxyphenyl)-2-methoxybenzamide
700573
Solid
306279-64-1
C16H17NO3
1g
CCOC1=CC=C(C=C1)NC(=O)C2=CC=CC=C2OC
271.316
271.32
Videos
SDS