missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-(4-Chlorophenyl)-5,6,7,8-tetrahydronaphthalene-1-carboxamide, 95%

Brand:  Alfa Aesar™ H65234.03

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15459486

  • 256.00 EUR / 1g


Estimated Shipment date
from Supplier 19-02-2019
Add to basket

Specifications

Chemical Identifiers

MFCD28098773

Specifications

N-(4-Chlorophenyl)-5,6,7,8-tetrahydronaphthalene-1-carboxamide
MFCD28098773
Solid
1g
285.77
95%
Videos
SDS