missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-[3-(Trifluoromethyl)phenyl]-2-methoxybenzamide, 97%

Brand:  Alfa Aesar™ H59888.03

Additional Details : CAS Number : 25331-53-7 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15485326

  • 646.00 EUR / 1g


Estimated Shipment date
from Supplier 02-02-2018
Add to basket

Specifications

Chemical Identifiers

N-[3-(Trifluoromethyl)phenyl]-2-methoxybenzamide
25331-53-7
295.26
MFCD00700882

Specifications

N-[3-(Trifluoromethyl)phenyl]-2-methoxybenzamide
25331-53-7
97%
MFCD00700882
1g
Solid
295.26
Videos
SDS