Learn More
Alfa Aesar™ N-(3-Methoxyphenyl)-5,6,7,8-tetrahydronaphthalene-1-carboxamide, 95%
Brand:  Alfa Aesar™ H65012.03
Additional Details : Weight : 0.00100kg
Quantity 1g
 View more versions of this product
Product Code. 15459316
  • 256.00 EUR / 1g
Estimated Shipment date
from Supplier 13-02-2019
Add to basket
Specifications
Chemical Identifiers
MFCD28098735
Specifications
N-(3-Methoxyphenyl)-5,6,7,8-tetrahydronaphthalene-1-carboxamide
MFCD28098735
Solid
1g
281.35
95%