missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-(3-Chloro-4-methylphenyl)-3-methylbenzamide, 97%

Brand:  Alfa Aesar™ H59343.03

Additional Details : CAS Number : 196700-88-6 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15485618

  • 646.00 EUR / 1g


Estimated Shipment date
from Supplier 05-02-2018
Add to basket

Specifications

Chemical Identifiers

N-(3-Chloro-4-methylphenyl)-3-methylbenzamide
196700-88-6
259.74
MFCD00088596

Specifications

N-(3-Chloro-4-methylphenyl)-3-methylbenzamide
Solid
259.74
1g
196700-88-6
97%
MFCD00088596
Videos
SDS