missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-(3,4-Difluorophenyl)-3,4-difluorobenzenesulfonamide, 97%

Brand:  Alfa Aesar™ H61583.03

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15446626

  • 720.00 EUR / 1g


Estimated Shipment date
from Supplier 19-06-2019
Add to basket

Specifications

Chemical Identifiers

MFCD01647053

Specifications

N-(3,4-Difluorophenyl)-3,4-difluorobenzenesulfonamide
MFCD01647053
Solid
1g
305.25
97%
Videos
SDS