missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-(3,4-Difluorophenyl)-2-methoxybenzamide, 97%

Brand:  Alfa Aesar™ H59367.03

Additional Details : CAS Number : 353782-02-2 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15445658

  • 646.00 EUR / 1g


Estimated Shipment date
from Supplier 24-04-2019
Add to basket

Specifications

Chemical Identifiers

353782-02-2
263.244
FRMKFKRUUVUISE-UHFFFAOYSA-N
871610
COC1=CC=CC=C1C(=O)NC2=CC(=C(C=C2)F)F
C14H11F2NO2
MFCD00981150
n-3,4-difluorophenyl-2-methoxybenzamide, cambridge id 5549150, n-3,4-difluorophenyl 2-methoxyphenyl carboxamide
N-(3,4-difluorophenyl)-2-methoxybenzamide

Specifications

N-(3,4-Difluorophenyl)-2-methoxybenzamide
353782-02-2
MFCD00981150
FRMKFKRUUVUISE-UHFFFAOYSA-N
N-(3,4-difluorophenyl)-2-methoxybenzamide
871610
Solid
1g
C14H11F2NO2
n-3,4-difluorophenyl-2-methoxybenzamide, cambridge id 5549150, n-3,4-difluorophenyl 2-methoxyphenyl carboxamide
COC1=CC=CC=C1C(=O)NC2=CC(=C(C=C2)F)F
263.244
263.24
97%
Videos
SDS