missing translation for 'onlineSavingsMsg'
Learn More

N-(2-Chloro-4-iodophenyl)-2-fluorobenzamide, 97%, Thermo Scientific™

Catalog Number p-7039783
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
433316-37-1
C13H8ClFINO
375.57
MFCD03240789
HMFPZLBLOKLAPU-UHFFFAOYSA-N
n-2-chloro-4-iodophenyl-2-fluorobenzamide, cambridge id 6920372
1324056
N-(2-chloro-4-iodophenyl)-2-fluorobenzamide
FC1=CC=CC=C1C(=O)NC1=C(Cl)C=C(I)C=C1
This item is not returnable. View return policy
433316-37-1
C13H8ClFINO
375.57
MFCD03240789
HMFPZLBLOKLAPU-UHFFFAOYSA-N
n-2-chloro-4-iodophenyl-2-fluorobenzamide, cambridge id 6920372
1324056
N-(2-chloro-4-iodophenyl)-2-fluorobenzamide
FC1=CC=CC=C1C(=O)NC1=C(Cl)C=C(I)C=C1

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 250 mg
Sensitivity Light sensitive
Formula Weight 375.57
Percent Purity 97%
Chemical Name or Material N-(2-Chloro-4-iodophenyl)-2-fluorobenzamide
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'