missing translation for 'onlineSavingsMsg'
Learn More

N-(2-Bromophenyl)-2,6-difluorobenzamide, 97%, Thermo Scientific™

Catalog Number 15435778
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
710291-06-8
C13H8BrF2NO
312.114
MFCD03551394
UQJVZAKVKZRKMF-UHFFFAOYSA-N
n-2-bromophenyl-2,6-difluorobenzamide, 2,6-difluorophenyl-n-2-bromophenyl carboxamide
4095566
N-(2-bromophenyl)-2,6-difluorobenzamide
C1=CC=C(C(=C1)NC(=O)C2=C(C=CC=C2F)F)Br
This item is not returnable. View return policy
710291-06-8
C13H8BrF2NO
312.114
MFCD03551394
UQJVZAKVKZRKMF-UHFFFAOYSA-N
n-2-bromophenyl-2,6-difluorobenzamide, 2,6-difluorophenyl-n-2-bromophenyl carboxamide
4095566
N-(2-bromophenyl)-2,6-difluorobenzamide
C1=CC=C(C(=C1)NC(=O)C2=C(C=CC=C2F)F)Br

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 250 mg
Beilstein 23653377
Formula Weight 312.11
Percent Purity 97%
Chemical Name or Material N-(2-Bromophenyl)-2,6-difluorobenzamide
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'