missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-(2,5-Dichlorophenyl)-2-methoxybenzamide, 97%

Brand:  Alfa Aesar™ H59658.03

Additional Details : CAS Number : 409348-29-4 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15486038

  • 646.00 EUR / 1g


Estimated Shipment date
from Supplier 31-08-2018
Add to basket

Specifications

Chemical Identifiers

409348-29-4
296.147
VFEAAVMXRIJMFE-UHFFFAOYSA-N
C14H11Cl2NO2
MFCD00751776
835335

Specifications

N-(2,5-Dichlorophenyl)-2-methoxybenzamide
97%
MFCD00751776
VFEAAVMXRIJMFE-UHFFFAOYSA-N
N-(2,5-dichlorophenyl)-2-methoxybenzamide
835335
Solid
409348-29-4
C14H11Cl2NO2
1g
COC1=CC=CC=C1C(=O)NC2=C(C=CC(=C2)Cl)Cl
296.147
296.15
Videos
SDS