missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Myc Epitope Tag Antibody (9E10), Alexa Fluor™ 350, Novus Biologicals™
Human Monoclonal Antibody
Brand: Bio-Techne NBP2-81018AF350
This item is not returnable.
View return policy
Description
Myc Epitope Tag Monoclonal antibody specifically detects Myc Epitope Tag in Epitope Tag samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen).
Specifications
Myc Epitope Tag | |
Monoclonal | |
Alexa Fluor 350 | |
50mM Sodium Borate | |
Human | |
Protein A purified | |
RUO | |
Epitope sequence, EQKLISEEDL, located in aa 410-419 of human c-myc protein. | |
Purified | |
Epitope Tag |
Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
90000000000 | |
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen | |
BHLHE39, Class E basic helix-loop-helix protein 39 (bHLHe39), Myc proto-oncogene protein, Proto-oncogene c-Myc, Transcription factor p64 | |
A synthetic peptide corresponding to aa 408-439 from C-terminus of human myc. | |
0.1 mL | |
Primary | |
Store at 4°C in the dark. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction