missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MRPL11 Monoclonal Antibody (OTI1E5), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA526102
Additional Details : Weight : 0.01000kg
Description
Mammalian mitochondrial ribosomal proteins are encoded by nuclear genes and help in protein synthesis within the mitochondrion. Mitochondrial ribosomes (mitoribosomes) consist of a small 28S subunit and a large 39S subunit. They have an estimated 75% protein to rRNA composition compared to prokaryotic ribosomes, where this ratio is reversed. Another difference between mammalian mitoribosomes and prokaryotic ribosomes is that the latter contain a 5S rRNA. Among different species, the proteins comprising the mitoribosome differ greatly in sequence, and sometimes in biochemical properties, which prevents easy recognition by sequence homology. This gene encodes a 39S subunit protein. Sequence analysis identified three transcript variants that encode different isoforms. Pseudogenes corresponding to this gene are found on chromosomes 5q and 12q.Specifications
MRPL11 | |
Monoclonal | |
1 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q9Y3B7 | |
MRPL11 | |
Full length human recombinant protein of MRPL11 produced in E.coli | |
100μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Immunohistochemistry (Paraffin), Western Blot | |
OTI1E5 | |
Unconjugated | |
MRPL11 | |
2410001P07Rik; 39S ribosomal protein L11, mitochondrial; CGI-113; cgi-113 protein; L11mt; mgc111024; Mitochondrial large ribosomal subunit protein uL11m; mitochondrial ribosomal protein L11; Mrpl11; MRP-L11 | |
Mouse | |
Affinity Chromatography | |
RUO | |
65003 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |