missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ Whole Serum, Deer, Purified, 2mL
Whole deer Serum with buffer salts
Brand: MP Biomedicals™ 0855994
Additional Details : Weight : 0.01000kg
Specifications
2° to 8°C | |
Deer Serum | |
0.2 μm filtration | |
Yes | |
59.3mg/mL |
Deer | |
2 mL | |
Serum Protein control in immunoelectrophoresis, Ouchterlony immunodiffusion, and other immunoprecipitation techniques, used as a blocking agent in fluorescence and enzyme immunoassays | |
7.3 |