Learn More
MP Biomedicals™ Protein G, Recombinant
Protein G is a monomeric protein lacking cysteine residues. Recombinant Protein G is a highly stable surface receptor that is capable of binding the Fc portion of immunoglobulins, especially IgGs from a large number of species.
Brand:  MP Biomedicals™ 08672651
Additional Details : Weight : 0.01000kg
Product Code. 11494525
  • 393.26 EUR / 1mg
Estimated Shipment: 09-04-2024
to see stock.
Add to basket
Description
Description
  • Protein G is a monomeric protein lacking cysteine residues.
  • Recombinant Protein G is a highly stable surface receptor that is capable of binding the Fc portion of immunoglobulins, especially IgGs from a large number of species.
  • This product can be used with immunologically active enzyme conjugates, ELISA, dot blot and Western Blot techniques, immunoprecipitation, and affinity purification of polyclonal and monoclonal antibodies.
  • Product is hygroscopic.
  • It is recommended to use the extinction coefficient for concentration calculations rather than weight.
Specifications
Specifications
ELISA, Immunoassays, Western Blotting
0°C to 5°C
1 mg
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Unless specified otherwise, for research or further manufacturing use only, not for direct human use.