Learn More
MP Biomedicals™ Protein G, Recombinant
Protein G is a monomeric protein lacking cysteine residues. Recombinant Protein G is a highly stable surface receptor that is capable of binding the Fc portion of immunoglobulins, especially IgGs from a large number of species.
Brand: MP Biomedicals™ 08672651
Additional Details : Weight : 0.01000kg
Description
- Protein G is a monomeric protein lacking cysteine residues.
- Recombinant Protein G is a highly stable surface receptor that is capable of binding the Fc portion of immunoglobulins, especially IgGs from a large number of species.
- This product can be used with immunologically active enzyme conjugates, ELISA, dot blot and Western Blot techniques, immunoprecipitation, and affinity purification of polyclonal and monoclonal antibodies.
- Product is hygroscopic.
- It is recommended to use the extinction coefficient for concentration calculations rather than weight.
Specifications
ELISA, Immunoassays, Western Blotting | |
0°C to 5°C | |
1 mg |
Unless specified otherwise, for research or further manufacturing use only, not for direct human use.