Learn More
MP Biomedicals™ Pepstatin A
Synthetic inhibitor for pepsin, renin, cathepsin D, and other acid proteases.
Brand: MP Biomedicals™ 0219536801
Additional Details : CAS Number : 26305-03-3 Weight : 0.01000kg
Description
Pepstatin A, [26305-03-3]; Isoval-Val-Val-Sta-Ala-Sta
C34H63N5O9, F.W. 685.9; Melting Point: 235-237°C
Specifications
Enzyme Assay, Inhibition Assay, Molecular Biology, Protein Purification, Sample Preparation | |
Pepstatin A | |
0°C |
685.91 | |
1mg | |
Powder |