Learn More
MP Biomedicals™ Pepstatin A
Synthetic inhibitor for pepsin, renin, cathepsin D, and other acid proteases.
Brand:  MP Biomedicals™ 0219536825
Additional Details : CAS Number : 26305-03-3 Weight : 0.00010kg
 View more versions of this product
Product Code. 11405332
  • 286.20 EUR / 25mg
Estimated Shipment: 17-05-2024
Description
Pepstatin A, [26305-03-3]; Isoval-Val-Val-Sta-Ala-Sta
C34H63N5O9, F.W. 685.9; Melting Point: 235-237°C
Specifications
Specifications
Enzyme Assay, Inhibition Assay, Molecular Biology, Protein Purification, Sample Preparation
Pepstatin A
0°C
685.91
25mg
Powder