missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Linbro™ PCR Rainbow Microcentrifuge Tubes

Use these V-shaped Plastic Snap Top Tubes for PCR applications

47.17 EUR - 89.89 EUR

Specifications

Capacity (Metric) 0.25 mL
Material Polypropylene
For Use With (Application) PCR applications
Includes Frosted writing area
Packaging 500 Case
View More Specs

Products 2
Product Code Brand Color Price Quantity & Availability  
Product Code Brand Color Price Quantity & Availability  
11431551
View Documents Promotion Details
MP Biomedicals™
0215950090
Natural This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
11441551
View Documents Promotion Details
MP Biomedicals™
0215950190
Assorted This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description

Description

Specifications

Specifications

0.25 mL
PCR applications
500 Case
Polypropylene
Frosted writing area
V
Videos
SDS
Documents

Documents

Product Certifications