Learn More
MP Biomedicals™ Linbro™ Microplate Sealer
For use with multi-well plates and tissue culture plates
65.99 EUR - 332.84 EUR
Specifications
Color | Clear |
---|---|
Material | Acetate |
Sterility | Non-sterile |
For Use With (Application) | Used for multi-well plates and tissue culture plates |
Product Type | Microplate Sealer |
Product Code | Brand | Material | No. per Pack | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Material | No. per Pack | Price | Quantity & Availability | ||||
11499565
|
MP Biomedicals™
097640105 |
Acetate | 100 |
65.99 EUR
Pack of 100 |
Estimated Shipment: 02-05-2024
Log in to see stock. |
||||
15702432
|
MP Biomedicals™
0976401C2 |
Acetate | 1000 |
332.84 EUR
Pack of 1000 |
Estimated Shipment: 02-05-2024
Log in to see stock. |
||||
Description
- Easy application and removal for all microplates for both short- and long-term storage Provide tight seal to minimize evaporation and condensation
- Suitable for use between -16 °C and 38 °C
- Transparent tape and is not pierceable
- Nonsterile
Specifications
Clear | |
Non-sterile | |
Microplate Sealer |
Acetate | |
Used for multi-well plates and tissue culture plates | |
-16°C to +38°C |