Learn More
MP Biomedicals™ Earle's Balanced Salts, 10X EBS Liq., Modified
10X EBS Liquid, Modified without sodium bicarbonate
Brand:  MP Biomedicals™ 091900154
Additional Details : Weight : 0.01000kg
Product Code. 11474835
  • 37.21 EUR / 500mL
Estimated Shipment: 14-06-2024
to see stock.
Specifications
Orange
10 X
500mL
4.44
15° to 30°C
Earle's Balanced Salt
Calcium Chloride Dihydrate 2649mg/L, Magnesium Sulfate 977mg/L, Potassium Chloride 4000mg/L, Sodium Chloride 68000mg/L, Sodium Phosphate Monobasic Monohydrate 1400mg/L, Dextrose 10000mg/L, Phenol Red Sodium Salt 170mg/L
Liquid