Learn More
MP Biomedicals™ CIRCLEGROW™ Large Capsules
Perfect for overnight or weekend cultures, CIRCLEGROW Medium is a super-rich bacterial growth medium that produces up to three times more plasmid DNA than traditional growth medium
237.44 EUR - 498.20 EUR
Specifications
Certifications/Compliance | Molecular biology Certified™ |
---|---|
Content And Storage | 15°C to 30°C |
Form | Dehydrated |
Grade | Molecular biology |
Product Type | Microbiology Media |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11333189
|
MP Biomedicals™
113000126 |
454 g |
237.44 EUR
454g |
Estimated Shipment: 11-06-2024
Log in to see stock. |
|||||
11363189
|
MP Biomedicals™
113000136 |
1000 g |
498.20 EUR
1kg |
Estimated Shipment: 11-06-2024
Log in to see stock. |
|||||
Description
CIRCLEGROW Medium consists of a protein and amino acid source, biological buffer, and a complex carbon source that gets metabolized at a much slower rate than glucose, thus prolonging the cell cycle and preventing cell death
during the log phase of growth. Cells grown with CIRCLEGROW Medium remain healthy for more than two days and continue to divide slowly after the log phase of the growth has been completed.
- Reduce hands-on work and preparation time
- Cultures do not need to be constantly monitored by O.D. readings due to the incremental post-log cell growth
- Obtain excellent yields of plasmid DNA from smaller culture volumes
- Confidently leave cultures overnight or over the weekend
- Capsules are the most convenient form of this unique growth media
- Pre-mixed CIRCLEGROW is also available in bulk powder, single-use pouches and sterile liquid
- Use 8 large capsules per liter;
- Contents/liter: Proprietary Formulation
Specifications
Molecular biology Certified™ | |
Dehydrated | |
Microbiology Media | |
For growth of Escherichia coli, plasmid and certified for plasmid and other bacterial cultures in liquid medium |
15°C to 30°C | |
Molecular biology | |
Yeast extract, Tryptone broth, Vitamins, Biological Buffers have been added for proper pH adjustment |