Learn More
MP Biomedicals™ Cellulase Y-C
From Trichoderma viride
Brand: MP Biomedicals™ 08320961
Additional Details : CAS Number : 9012-54-8 Weight : 0.01000kg
Description
- Very high filter paper decomposing activity
- Appreciable xylanase and hemicellulase activity
- For use with pectolyase Y-23 for plant cell wall removal
Specifications
16,000 U/g | |
4 to 5 | |
2°C to 8°C | |
10 g | |
Solid |
Light Brown | |
Removed cell walls from plant tissues in shorter incubation period without loss of biological activity of the materials for protoplast preparation | |
Trichoderma viride | |
Cellulase Y-C |