Learn More
MP Biomedicals™ Albumin, Bovine, Fatty Acid-Free
Prepared by Fraction V method
Brand: MP Biomedicals™ 0215240110
Additional Details : CAS Number : 9048-46-8 Weight : 1.36080kg
Description
- Low fatty acids
- Purity: >98%
- pH: 7
For best cell culture performance, see alternate Mfr. No. 180575
Specifications
Immunohematology, Serology | |
Bovine Albumin | |
10g | |
2° to 8°C |
66.338 | |
5.2 to 7 | |
Bovine | |
Powder |