Learn More
Mouse IgG1 kappa Isotype Control (P3.6.2.8.1), Super Bright™ 436, eBioscience™, Invitrogen™
Mouse Isotype Control
146.00 EUR - 308.00 EUR
Specifications
Antigen | Mouse IgG1 kappa |
---|---|
Clone | P3.6.2.8.1 |
Concentration | 0.2 mg/mL |
Applications | Control, Flow Cytometry |
Conjugate | Super Bright 436 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15815533
|
Affymetrix eBioscience
62-4714-80 |
25 μg |
146.00 EUR
25µg |
Estimated Shipment: 24-05-2024
Log in to see stock. |
|||||
16311264
|
Affymetrix eBioscience
62-4714-82 |
100 μg |
308.00 EUR
100µg |
Estimated Shipment: 24-05-2024
Log in to see stock. |
|||||
Description
Description: The monoclonal mouse IgG1 K immunoglobulin is useful as an isotype control. Applications Reported: This P3.6.2.8.1 antibody has been reported for use in flow cytometric analysis. Applications Tested: This P3.6.2.8.1 antibody has been tested by flow cytometric analysis of normal human peripheral blood cells and mouse splenocytes. Use the isotype control at the same concentration as the experimental antibody. Super Bright 436 can be excited with the violet laser line (405 nm) and emits at 436 nm. We recommend using a 450/50 bandpass filter, or equivalent. Please make sure that your instrument is capable of detecting this fluorochrome. When using two or more Super Bright dye-conjugated antibodies in a staining panel, it is recommended to use Super Bright Complete Staining Buffer (Product # SB-4401) to minimize any non-specific polymer interactions. Please refer to the datasheet for Super Bright Staining Buffer for more information. Excitation: 405 nm; Emission: 436 nm; Laser: Violet Laser Super Bright Polymer Dyes are sold under license from Becton, Dickinson and Company.
Mouse IgG1 is a subtype of immunoglobulin G antibody that is produced by B cells in response to an antigen. It is the most abundant IgG subtype in the serum and plays a crucial role in the adaptive immune response. Mouse IgG1 is composed of two heavy chains and two light chains, and it is involved in opsonization, complement activation, and antibody-dependent cell-mediated cytotoxicity. It is also involved in the regulation of immune responses, including the activation of T cells and the inhibition of B cell activation. Mouse IgG1 is widely used in research and diagnostic applications, including ELISA, Western blotting, and immunohistochemistry. It is also used in therapeutic applications, such as immunotherapy and vaccine development.Specifications
Mouse IgG1 kappa | |
0.2 mg/mL | |
Super Bright 436 | |
Mouse | |
Not Applicable | |
IgG1; Igh-4; Ighg1; Immunoglobulin G; Immunoglobulin G1; immunoglobulin heavy constant gamma 1 (G1m marker); ImmunoglobulinG; ImmunoglobulinG1; VH7183 | |
Isotype Control (Primary) | |
4° C, store in dark, DO NOT FREEZE! |
P3.6.2.8.1 | |
Control, Flow Cytometry | |
Liquid | |
RUO | |
PBS with BSA and 0.09% sodium azide; pH 7.2 | |
IgG1 κ | |
Affinity chromatography |
For Research Use Only.