Learn More
Mouse IgG1 Isotype Control (MOPC-21), DyLight™ 488, Invitrogen™
Mouse Isotype Control
Brand: Invitrogen MA1191D488
Additional Details : Weight : 0.00900kg
Description
MA1-191-D488 can be used as a negative control in any immunoassay where a DyLight 488-conjugated primary antibody of the mouse IgG1 isotype is used. DyLight 488 has an excitation/emission of 493/518 nm. MA1-191-D488 can be used as isotype control for Dylight 488 conjugated IgG1 mouse monoclonal antibodies in immunofluorescence and flow cytometry analysis.
Mouse IgG1 Control antibody specifically detects Mouse IgG1 in Not Applicable samples. It is validated for Control, Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot.Specifications
Mouse IgG1 | |
MOPC-21 | |
DyLight 488 | |
IgG1; Igh-4; Ighg1; Immunoglobulin G; Immunoglobulin G1; immunoglobulin heavy constant gamma 1 (G1m marker); ImmunoglobulinG; ImmunoglobulinG1; VH7183 | |
Size-exclusion chromatography | |
RUO | |
Not Applicable | |
Liquid |
Control, Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot | |
0.1 mg/mL | |
PBS with 0.2% BSA and 15mM sodium azide; pH 7.4 | |
Mouse | |
100 μg | |
Isotype Control (Primary) | |
4° C, do not freeze | |
IgG1 κ |