Learn More
SERPINA1 Mouse anti-Human, Monoclonal Antibody, Abnova™
Mouse monoclonal antibody raised against native SERPINA1.
Brand: Abnova MAB6142.100ug
Additional Details : Weight : 0.02000kg
Description
The protein encoded by this gene is secreted and is a serine protease inhibitor whose targets include elastase, plasmin, thrombin, trypsin, chymotrypsin, and plasminogen activator. Defects in this gene can cause emphysema or liver disease. Several transcript variants encoding the same protein have been found for this gene. [provided by RefSeq
Specifications
SERPINA1 | |
Monoclonal | |
Mouse monoclonal antibody raised against native SERPINA1. | |
In PBS, pH 7.4 | |
A1A/A1AT/AAT/MGC23330/MGC9222/PI/PI1/PRO2275 | |
Mouse | |
100 μg | |
Primary | |
These antibodies specifically recognize alpha-1-antichymotrypsin. They do not cross-react with alpha-1-antitrypsin. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. |
|
IgG1 |
ELISA, Immunofluorescence | |
Unconjugated | |
The optimal working dilution should be determined by the end user. | |
SERPINA1 | |
SERPINA1 | |
Native purified SERPINA1 from human serum. | |
RUO | |
5265 | |
Human | |
Liquid |