Learn More
CD34, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant CD34.
Brand: Abnova H00000947-A01.50uL
Additional Details : Weight : 0.00010kg
Description
CD34 is a monomeric cell surface antigen with a molecular mass of approximately 110kDa that is selectively expressed on human hematopoietic progenitor cells.[supplied by OMIM
Sequence: SLDNNGTATPELPTQGTFSNVSTNVSYQETTTPSTLGSTSLHPVSQHGNEATTNITETTVKFTSTSVITSVYGNTNSSVQSQTSVISTVFTTPANVSTPETTLKPSLSPGSpecifications
CD34 | |
Polyclonal | |
Mouse polyclonal antibody raised against a partial recombinant CD34. | |
CD34 | |
CD34 | |
CD34 (NP_001764, 32 a.a. to 141 a.a) partial recombinant protein with GST tag. | |
RUO | |
947 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
ELISA, Western Blot | |
Unconjugated | |
50% glycerol | |
NM_001773 | |
Mouse | |
50 μL | |
Primary | |
Human | |
Antisera |
For Research Use Only