Learn More
CD200R1, Mouse, Clone: OX108, Abnova™
Mouse monoclonal antibody raised against recombinant CD200R1.
Brand: Abnova MAB5234.200ug
Additional Details : Weight : 0.02000kg
Description
This gene encodes a receptor for the OX-2 membrane glycoprotein. Both the receptor and substrate are cell surface glycoproteins containing two immunoglobulin-like domains. This receptor is restricted to the surfaces of myeloid lineage cells and the receptor-substrate interaction may function as a myeloid downregulatory signal. Mouse studies of a related gene suggest that this interaction may control myeloid function in a tissue-specific manner. Alternative splicing of this gene results in multiple transcript variants. [provided by RefSeq
Specifications
CD200R1 | |
Monoclonal | |
Unconjugated | |
Flow Cytometry (10ul of the suggested working dilution (1:50-1:100) to label 106 cells in 100ul) The optimal working dilution should be determined by the end user. | |
CD200R1 | |
CD200R1 | |
Recombinant fusion protein corresponding to human CD200R1. | |
RUO | |
131450 | |
Human | |
Liquid |
Flow Cytometry | |
OX108 | |
Mouse monoclonal antibody raised against recombinant CD200R1. | |
In PBS, pH 7.4 (0.09% sodium azide) | |
CD200R/HCRTR2/MOX2R/OX2R | |
Mouse | |
200 μg | |
Primary | |
This antibody recognizes human CD200R, a cell surface glycoprotein (also known as OX2R). | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
IgG1 |