Learn More
Invitrogen™ Mouse ADAMTS4 (aa 394-403) Synthetic Peptide
Synthetic Peptide
Brand: Invitrogen™ PEP234
Additional Details : CAS Number : 69-74-9 Weight : 0.01000kg
Description
Mouse ADAMTS4 (residues 394-403) Synthetic Peptide, PEP-234, from Invitrogen.
PEP-234 is a 11 amino acid synthetic peptide corresponding to amino acid residues 394-403 from mouse ADAMTS4. The sequence of this peptide is (amino to carboxy terminus): C V (394) M A H V D P E E P (403).This peptide may be used for neutralization and control experiments with the polyclonal antibody that reacts with this product, catalog # PA1-1749A. Using a solution of peptide of equal volume and concentration to the corresponding antibody will yield a large molar excess of peptide (∼70-fold) for competitive inhibition of antibody-protein binding reactions.Reconstitute with 0.1 ml of distilled water.Specifications
240913 | |
purified | |
Mouse ADAMTS4 (aa 394-403) | |
-20°C. Avoid freeze-thaw cycles. | |
Lyophilized | |
Control | |
Yes | |
Peptide | |
RUO - research use only |
Mouse | |
Q8BNJ2 | |
0.5 mg/mL | |
Synthetic Peptide | |
peptide with no preservative | |
Adamts4 | |
No preservative | |
50 μg | |
C V(394) M A H V D P E E P(403) |