Learn More
SCHOTT AG Lighting and Imaging™ Mounting Bracket for EasyLED Spotlight Plus and KL300LED
The Mounting Bracket allows ergonomic operation of Easy LED Spotlight and KL300LED. Connect directly to the microscope via column-adapter.
32.35 EUR - 45.45 EUR
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
11796718
|
SCHOTT AG Lighting and Imaging™
120225 |
25 mm dia. Column |
32.35 EUR
Each |
Estimated Shipment: 27-06-2024
Log in to see stock. |
|||||
15713012
|
SCHOTT AG Lighting and Imaging™
120229 |
30 mm dia. Column |
32.65 EUR
1 unit |
Estimated Shipment: 30-07-2024
Log in to see stock. |
|||||
11756728
|
SCHOTT AG Lighting and Imaging™
120232 |
32 mm dia. Column |
32.35 EUR
Each |
Estimated Shipment: 04-06-2024
Log in to see stock. |
|||||
15723012
|
SCHOTT AG Lighting and Imaging™
120240 |
Zeiss Stand K, Stand M |
32.65 EUR
1 unit |
Estimated Shipment: 30-07-2024
Log in to see stock. |
|||||
11358330
|
SCHOTT AG Lighting and Imaging™
120250 |
Base 158.340 |
45.45 EUR
Each |
Estimated Shipment: 28-05-2024
Log in to see stock. |
|||||
Description
- Connects the EasyLED Spotlight Plus or KL300LED to the microscope column, base or a base.
Specifications
Stereo Microscopy Illumination |