Learn More
Uvex™ Motion S2 SRC Perforated Safety Shoes
Extremely lightweight safety shoes with a sporty design constructed from synthetic materials, making them suitable for those who are allergic to chrome. Protection class S2 in accordance with EN ISO 20345:2011 with additional marking for excellent slip resistance (SRC).
140.00 EUR
Specifications
Color | Black, Silver |
---|---|
Liner Material | Distance Mesh |
Material | Microvelour |
Sole Material | Dual Density Polyurethane (PU) |
Toe Material | Steel |
Product Code | Brand | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | |||||
15308262
|
Uvex™
6978836 |
36 |
140.00 EUR
Each |
Estimated Shipment: 30-05-2024
Log in to see stock. |
|||||
15318262
|
Uvex™
6978837 |
37 |
140.00 EUR
Each |
Estimated Shipment: 30-05-2024
Log in to see stock. |
|||||
15328262
|
Uvex™
6978838 |
38 |
140.00 EUR
Each |
Estimated Shipment: 30-05-2024
Log in to see stock. |
|||||
15338262
|
Uvex™
6978839 |
39 |
140.00 EUR
Each |
Estimated Shipment: 30-05-2024
Log in to see stock. |
|||||
15348262
|
Uvex™
6978840 |
40 |
140.00 EUR
Each |
Estimated Shipment: 30-05-2024
Log in to see stock. |
|||||
15358262
|
Uvex™
6978841 |
41 |
140.00 EUR
Each |
Estimated Shipment: 30-05-2024
Log in to see stock. |
|||||
15388262
|
Uvex™
6978844 |
44 |
140.00 EUR
Each |
Estimated Shipment: 30-05-2024
Log in to see stock. |
|||||
15308272
|
Uvex™
6978846 |
46 |
140.00 EUR
Each |
Estimated Shipment: 30-05-2024
Log in to see stock. |
|||||
15318272
|
Uvex™
6978847 |
47 |
140.00 EUR
Each |
Estimated Shipment: 30-05-2024
Log in to see stock. |
|||||
15328272
|
Uvex™
6978848 |
48 |
140.00 EUR
Each |
Estimated Shipment: 30-05-2024
Log in to see stock. |
|||||
Description
- Lightweight S2 safety shoes with a fashionable, sporty design
- Made from synthetic materials, making them suitable for those who are allergic to chrome
- Protection class S2 in accordance with EN ISO 20345:2011, additional marking for excellent slip resistance (SRC)
- Meets the ESD specification with bleeder resistor under 35 megaohms
- Self-cleaning, lightweight PU outsole with optimal abrasion resistance, shock absorption, and excellent slip resistance
- Steel toe cap with a stabilizing anti-twist heel cap to prevent ankle twisting
- All soles are free of silicones, plasticizers, and other paint wetting impairment substances
- Anti-static Uvex 3D Hydroflex™ foam insole for optimal shock absorption around heel and forefoot
- Elastic laces allow for quick, individual adjustment
- Suitable for use with orthopaedic adjustments and insoles
- Ideal foot climate constantly maintained thanks to Uvex Climazone
Specifications
Black, Silver | |
Microvelour | |
Steel | |
Men's | |
S2 |
Distance Mesh | |
Dual Density Polyurethane (PU) | |
11 cm | |
Motion Style S2 SRC |