missing translation for 'onlineSavingsMsg'
Learn More

Molybdenum(VI) oxide, ≥99.5%, Reagent Grade, Honeywell™

Catalog Number 15667920
Click to view available options
Quantity:
100 g
500 g
1 kg
Packaging:
Poly bottle
This item is not returnable. View return policy
1313-27-5
MoO3
143.95
MFCD00003469
JKQOBWVOAYFWKG-UHFFFAOYSA-N
molybdenum trioxide, molybdenum vi oxide, molybdic anhydride, molybdic trioxide, molybdena, molybdic oxide, molybdenum oxide, molybdic acid anhydride, molybdenum peroxide
14802
CHEBI:30627
trioxomolybdenum
O=[Mo](=O)=O
This item is not returnable. View return policy
1313-27-5
MoO3
143.95
MFCD00003469
JKQOBWVOAYFWKG-UHFFFAOYSA-N
molybdenum trioxide, molybdenum vi oxide, molybdic anhydride, molybdic trioxide, molybdena, molybdic oxide, molybdenum oxide, molybdic acid anhydride, molybdenum peroxide
14802
CHEBI:30627
trioxomolybdenum
O=[Mo](=O)=O

Reagent Grade,≥99.5%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY
Melting Point 795°C
Color Gray
Physical Form Powder
Quantity 1 kg
Linear Formula MoO3
UN Number NONH for all modes of transport
Formula Weight 143.94g/mol
Percent Purity ≥99.5%
Odor Odorless
Packaging Poly bottle
Chemical Name or Material Molybdenum(VI) oxide
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'