Learn More
Molecular Sieve Uop Type 3A, Honeywell Fluka™
Beads
Brand: Honeywell Fluka 02573-1KG
Additional Details : CAS Number : 1318-02-1 Weight : 1.00000kg
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
1 kg | |
Molecular Sieve Uop Type 3A | |
1318-02-1 | |
NONH for all modes of transport |