Learn More
Invitrogen™ NeuroTrace™ Multicolor Tissue-Labeling Kit - DiO, DiI, DiD pastes, 500 mg each
Contains one paste each of DiI, DiO or DiD mixed into an inert, water-resistant gel
Brand: Invitrogen™ N22884
817.00 EUR valid until 2024-05-31
Use promo code "21664" to get your promotional price.
Additional Details : Weight : 0.00525kg
Description
- Ready to use as supplied and can be applied directly to live or fixed tissue specimens using the tip of a needle
- Improves penetration of dye into bundled neurons, labeling axons both on and below surface
Cell Analysis, Cell Tracing and Tracking, Cellular Imaging, Immunofluorescence (IF), Immunofluorescence Staining and Detection, Neuronal Tracing
Order Info
Shipping Condition: Room temperature
Specifications
Green, Yellow, Red | |
Tissue Labeling Kit | |
Room Temperature | |
Cell Membranes & Lipids |
Fluorescent | |
Store at room temperature and protect from light. | |
Fluorescence Microscope | |
NEUROTRACE™ |
For Research Use Only. Not for use in diagnostic procedures.