Learn More
Miele Professional™ Mobile Injector, Module A 301/1
Module A 301/1 for the optimum loading of laboratory glassware
Brand:  Miele Professional™ 11116950
Additional Details : Weight : 2.75400kg
Product Code. 15435021
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
  • Suitable for laboratory glassware such as Erlenmeyer flasks
  • equipped with 18 injector nozzles E 351 (4 x 160 mm)
  • 16 injector nozzles locking mechanisms for nozzle E 353
  • Nozzle spacing 78 mm across the width, 84 mm across the depth