Learn More
MMP3 Monoclonal Antibody (SL-1 ID3), Biotin, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA514199
Additional Details : Weight : 0.01000kg
Description
MA5-14199 targets MMP-3 (Stromelysin-1) in IF and WB applications and shows reactivity with Human samples. The MA5-14199 immunogen is aPMA (4-Aminophenylmercuric acetate) activated Human stromelysin-1 (SL-1).
MMP3 (stromelysin-1) belongs to the peptidase M10A subfamily. It is secreted in its pro-protein form, and is activated by extracellular proteinase cleavage. MMP3 is capable of binding to zinc and calcium ions, and has metallo-endopeptidase activity. The active enzyme degrades collagen, gelatin, fibronectin, and laminin. It is involved in normal cellular processes such as wound repair and tissue remodeling. Mutations in the gene can result in coronary heart disease 6.Specifications
MMP3 | |
Monoclonal | |
0.2 mg/mL | |
PBS with 0.2% BSA and 0.09% sodium azide; pH 7.4 | |
P08254 | |
MMP3 | |
APMA (4-Aminophenylmercuric acetate) activated Human stromelysin-1 (SL-1) | |
500 μL | |
Primary | |
Human | |
Antibody | |
IgG1 κ |
Immunocytochemistry, Western Blot | |
SL-1 ID3 | |
Biotin | |
MMP3 | |
CHDS6; EC 3.4.24.17; EMS-2; matrix metallo protease; matrix metallopeptidase 3; matrix metallopeptidase 3 (stromelysin 1, progelatinase); matrix metalloproteinase 3; matrix metalloproteinase 3 (stromelysin 1, progelatinase); matrix metalloproteinase 3 precursor; matrix metalloproteinase-3; metalloproteinase 3 receptor; MMP; MMP10; MMP3; MMP-3; MMP-3 protein; MMPs; progelatinase; proteoglycanase; SL-1; SLN1; SLN-1; STMY; STMY1; Str1; STR-1; stromelysin; stromelysin 1; Stromelysin1; stromelysin-1; Transin1; Transin-1 | |
Mouse | |
Protein G | |
RUO | |
4314 | |
4° C | |
Liquid |