Learn More
HUBER Minichiller 300
The compact Minichiller is an economic and environmentally friendly cooling solution for many laboratory applications. Minichiller can save up to 48,000 l of water in a working week and allow stable application conditions due to constant values for temperature, pressure and through flow quantities.
Brand:  HUBER 3006.0063.99
Additional Details : Weight : 46.00000kg
Product Code. 15565794
  • 3641.10 EUR / Each
Estimated Shipment: 26-06-2024
Description
  • High efficiency
  • Precise temperature control
  • Powerful circulation pump
  • Robust stainless construction
  • Reliable continuously operation with warning functions
  • Easy to fill and empty
  • Illuminated filling level indicator
  • LED temperature display