Learn More
Cytiva (formerly Pall Lab) Mini Profile™ Capsules with Profile Star Filter
Mini Profile™ Capsules designed for research applications only.
Brand: Cytiva (formerly Pall Lab) 12070
Additional Details : Weight : 0.39900kg
Description
- Users have the convenient choice between high void-volume depths medium or high-area pleated depth filter
- Modules can come in different filter specifications to meet the specific needs of an early phase process development
Specifications
Spun Polypropylene | |
Non-sterile | |
3/Pk. |
1.5 μm | |
90 cm2 | |
9/16 in. Hose Barb |