missing translation for 'onlineSavingsMsg'
Learn More

Millesimal Calibration Buffer pH 1.679, Hanna Instruments™

Catalog Number 11606990
Click to view available options
Quantity:
500 mL
This item is not returnable. View return policy
This item is not returnable. View return policy
Color Colorless
pH 1.679
Physical Form Liquid
Quantity 500 mL
Solution Type Buffer
Formulation >1% to <5% Potassium Tetraoxalate Dihydrate
Odor Odorless
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'