Learn More
Kinesis™ Microwave Vials
Brand:  Kinesis™ VMW05-C-100-SB
Additional Details : Weight : 0.50000kg
Product Code. 15198102
  • 419.00 EUR / Pack of 100
Estimated Shipment: 10-09-2024
Description
KX, for use with Initiator Systems (EXP only) from Biotage, contamination free 1st class hydrolytic borosilicate glass, resealable septa, 40°C to 300°C, 2.0mL to 5.0mL