Learn More
Thermo Scientific™ Microscope Slides, Cut, 1mm
Premium slides are manufactured through a unique proprietary cleaning process intended to provide consistent results and ease interpretation. This process allows these slides to to be used in routine histology and microbiology applications; and specifically in applications by delivering consistently crisp, featherededge blood smears with good morphology.
Brand:  Thermo Scientific™ AAAC0170076002EA0MNZ#0#G#
Additional Details : Weight : 1.00000kg
 View more versions of this product
Product Code. 15284297
  • 159.84 EUR / Pack of 50
Call for latest availability
+358 9 8027 6280
Add to basket
Description
Description
  • Manufactured from Swiss extra-white soda-lime glass
  • Reduced iron content for low self-fluorescence
  • Greenish shimmer minimized
  • Guaranteed optimum, faultless transparency
  • Clean, flat and suitable for immediate use
Specifications
Specifications
76mm
1mm
Soda-lime glass
No
17mm
Uncoated
50 pieces in paper
76 x 17mm
Videos